(2-Bromo-6-fluoro-3-propoxyphenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

318599-1G 318599-500MG 318599-5G
77469-052EA 415.09 USD
77469-052 77481-728 77492-122
(2-Bromo-6-fluoro-3-propoxyphenyl)(m-tolyl)methanol ≥95%
(2-Bromo-6-fluoro-3-propoxyphenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₇H₁₈BrFO₂
Storage Temperature: Ambient
CAS Number: 3002436-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR