1-(2-Bromo-6-fluoro-3-propoxyphenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

318596-1G 318596-500MG 318596-5G
77469-048EA 415.09 USD
77469-048 77481-724 77492-118
1-(2-Bromo-6-fluoro-3-propoxyphenyl)prop-2-en-1-ol ≥95%
1-(2-Bromo-6-fluoro-3-propoxyphenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₁₂H₁₄BrFO₂
MW: 289.1407 g/mol
Storage Temperature: Ambient
CAS Number: 3002435-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR