2-Bromo-4-fluoro-3-iodo-1-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

318590-1G 318590-500MG 318590-5G
77469-044EA 415.09 USD
77469-044 77481-720 77492-114
2-Bromo-4-fluoro-3-iodo-1-propoxybenzene ≥95%
2-Bromo-4-fluoro-3-iodo-1-propoxybenzene

Precaución: For research use only

Formula: C₉H₉BrFIO
MW: 358.9740 g/mol
Storage Temperature: Ambient
CAS Number: 3002500-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR