(2,6-Difluoro-4-methoxyphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

318579-1G 318579-500MG 318579-5G
77469-040EA 415.09 USD
77469-040 77481-716 77492-110
(2,6-Difluoro-4-methoxyphenyl)triethylsilane ≥95%
(2,6-Difluoro-4-methoxyphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₂₀F₂OSi
MW: 258.3796 g/mol
Storage Temperature: Ambient
CAS Number: 3002496-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR