(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(butyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

318576-1G 318576-500MG 318576-5G
77469-036EA 415.09 USD
77469-036 77481-712 77492-106
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(butyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(butyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₁BrF₄S
Storage Temperature: Ambient
CAS Number: 3002500-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR