1-Bromo-2,4,5-trimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

31844-1G 31844-500MG 31844-5G
77469-032EA 78.28 USD
77469-032 77481-708 77492-102
1-Bromo-2,4,5-trimethoxybenzene ≥97%
1-Bromo-2,4,5-trimethoxybenzene

Precaución: For research use only

Formula: C₉H₁₁BrO₃
MW: 247.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094684
CAS Number: 20129-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR