1-Bromo-3-n-butyloxy-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

31794-1G 31794-500MG 31794-5G
77469-028EA 197.76 USD
77469-028 77481-704 77492-098
1-Bromo-3-n-butyloxy-4-fluorobenzene ≥95%
1-Bromo-3-n-butyloxy-4-fluorobenzene

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917772
CAS Number: 1309933-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR