1-Bromo-2,4-difluoro-5-ethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

31777-1G 31777-500MG 31777-5G
77469-024EA 283.25 USD
77469-024 77481-700 77492-094
1-Bromo-2,4-difluoro-5-ethoxybenzene ≥95%
1-Bromo-2,4-difluoro-5-ethoxybenzene

Precaución: For research use only

Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD27931838
CAS Number: 900175-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR