(2-Bromo-3-chloro-6-fluorophenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

316746-1G 316746-500MG 316746-5G
77469-020EA 415.09 USD
77469-020 77481-696 77492-090
(2-Bromo-3-chloro-6-fluorophenyl)(m-tolyl)methanol ≥95%
(2-Bromo-3-chloro-6-fluorophenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₁BrClFO
MW: 329.5919 g/mol
Storage Temperature: Ambient
CAS Number: 3002482-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR