(2-Bromo-3-chloro-6-fluorophenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

316743-1G 316743-500MG 316743-5G
77469-016EA 415.09 USD
77469-016 77481-692 77492-086
(2-Bromo-3-chloro-6-fluorophenyl)(propyl)sulfane ≥95%
(2-Bromo-3-chloro-6-fluorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₉BrClFS
Storage Temperature: Ambient
CAS Number: 3002513-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR