1-(2-Bromo-3-chloro-6-fluorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

316741-1G 316741-500MG 316741-5G
77469-012EA 415.09 USD
77469-012 77481-688 77492-082
1-(2-Bromo-3-chloro-6-fluorophenyl)propan-1-ol ≥95%
1-(2-Bromo-3-chloro-6-fluorophenyl)propan-1-ol

Precaución: For research use only

Formula: C₉H₉BrClFO
MW: 267.5225 g/mol
Storage Temperature: Ambient
CAS Number: 3002496-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR