(2-Bromo-3-chloro-6-fluorophenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

316736-1G 316736-500MG 316736-5G
77469-004EA 415.09 USD
77469-004 77481-680 77492-074
(2-Bromo-3-chloro-6-fluorophenyl)(4-ethylphenyl)methanol ≥95%
(2-Bromo-3-chloro-6-fluorophenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₃BrClFO
MW: 343.6185 g/mol
Storage Temperature: Ambient
CAS Number: 3002543-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR