1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

316702-1G 316702-500MG 316702-5G
77469-000EA 415.09 USD
77469-000 77481-674 77492-068
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₆H₉BrF₆O₂
Storage Temperature: Ambient
CAS Number: 3002483-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR