1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

316701-1G 316701-500MG 316701-5G
77468-996EA 415.09 USD
77468-996 77481-672 77492-066
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2-methylbutan-1-ol ≥95%
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₈H₂₀BrFO₂
MW: 367.2526 g/mol
Storage Temperature: Ambient
CAS Number: 3002469-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR