4-Bromo-2-(isopropylthio)benzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

316683-1G 316683-500MG 316683-5G
77468-992EA 415.09 USD
77468-992 77481-668 77492-062
4-Bromo-2-(isopropylthio)benzo[b]thiophene ≥95%
4-Bromo-2-(isopropylthio)benzo[b]thiophene

Precaución: For research use only

Formula: C₁₁H₁₁BrS₂
MW: 287.2390 g/mol
Storage Temperature: Ambient
CAS Number: 3002483-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR