1-[3-[(2-Fluorophenyl)methoxy]phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

316563-1G 316563-500MG 316563-5G
77468-988EA 538.69 USD
77468-988 77481-664 77492-058
1-[3-[(2-Fluorophenyl)methoxy]phenyl]ethanone ≥95%
1-[3-[(2-Fluorophenyl)methoxy]phenyl]ethanone

Precaución: For research use only

Formula: C₁₅H₁₃FO₂
MW: 244.26 g/mol
Storage Temperature: Ambient
CAS Number: 478159-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR