1-[3-[(2-Methylphenyl)methoxy]phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

316244-1G 316244-500MG 316244-5G
77468-984EA 538.69 USD
77468-984 77481-660 77492-054
1-[3-[(2-Methylphenyl)methoxy]phenyl]ethanone ≥95%
1-[3-[(2-Methylphenyl)methoxy]phenyl]ethanone

Precaución: For research use only

Formula: C₁₆H₁₆O₂
MW: 240.3 g/mol
Storage Temperature: Ambient
CAS Number: 312592-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR