4-Butyl-4'-(methylthio)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

315283-1G 315283-500MG 315283-5G
77468-980EA 415.09 USD
77468-980 77481-656 77492-048
4-Butyl-4'-(methylthio)-1,1'-biphenyl ≥95%
4-Butyl-4'-(methylthio)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₇H₂₀S
MW: 256.41 g/mol
Storage Temperature: Ambient
CAS Number: 104560-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR