4'-Butyl[1,1'-biphenyl]-4-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

315247-1G 315247-500MG 315247-5G
77468-976EA 883.74 USD
77468-976 77481-652 77492-044
4'-Butyl[1,1'-biphenyl]-4-carboxaldehyde ≥95%
4'-Butyl[1,1'-biphenyl]-4-carboxaldehyde

Precaución: For research use only

Formula: C₁₇H₁₈O
MW: 238.32 g/mol
Storage Temperature: Ambient
CAS Number: 93972-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR