1-(Benzyloxy)-3-fluoro-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

31453-1G 31453-5G
77468-968EA 62.83 USD
77468-968 77492-036
1-(Benzyloxy)-3-fluoro-5-(trifluoromethyl)benzene ≥97%
1-(Benzyloxy)-3-fluoro-5-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₄H₁₀F₄O
MW: 270.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564020
CAS Number: 1807772-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR