(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

314287-1G 314287-500MG 314287-5G
77468-960EA 415.09 USD
77468-960 77481-642 77492-028
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(cyclohexyl)methanol ≥95%
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₇BrClFO
MW: 335.6395 g/mol
Storage Temperature: Ambient
CAS Number: 3002527-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR