(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

314284-1G 314284-500MG 314284-5G
77468-956EA 415.09 USD
77468-956 77481-638 77492-024
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)triethylsilane ≥95%
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₁₉BrClFSi
MW: 337.7308 g/mol
Storage Temperature: Ambient
CAS Number: 3002435-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR