1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

314281-1G 314281-500MG 314281-5G
77468-952EA 415.09 USD
77468-952 77481-634 77492-020
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-methylpropan-1-ol ≥95%
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃BrClFO
MW: 295.5757 g/mol
Storage Temperature: Ambient
CAS Number: 3002480-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR