1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

314279-1G 314279-500MG 314279-5G
77468-948EA 415.09 USD
77468-948 77481-630 77492-016
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₅BrClFO
MW: 309.6023 g/mol
Storage Temperature: Ambient
CAS Number: 3002499-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR