1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

314277-1G 314277-500MG 314277-5G
77468-944EA 415.09 USD
77468-944 77481-626 77492-012
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₄BrCl₂F₃O
MW: 335.9326 g/mol
Storage Temperature: Ambient
CAS Number: 3002473-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR