2-Bromo-3-chloro-6-fluoro-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

314275-1G 314275-500MG 314275-5G
77468-940EA 415.09 USD
77468-940 77481-622 77492-008
2-Bromo-3-chloro-6-fluoro-4-methylbenzoic acid ≥95%
2-Bromo-3-chloro-6-fluoro-4-methylbenzoic acid

Precaución: For research use only

Formula: C₈H₅BrClFO₂
Storage Temperature: Ambient
CAS Number: 2937598-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR