1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

314272-1G 314272-500MG 314272-5G
77468-936EA 415.09 USD
77468-936 77481-618 77492-004
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2-difluoroethanone ≥95%
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O
MW: 301.4876 g/mol
Storage Temperature: Ambient
CAS Number: 3002435-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR