(6-Bromo-2-fluoro-3-propoxyphenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

314269-1G 314269-500MG 314269-5G
77468-932EA 415.09 USD
77468-932 77481-614 77492-000
(6-Bromo-2-fluoro-3-propoxyphenyl)(m-tolyl)methanol ≥95%
(6-Bromo-2-fluoro-3-propoxyphenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₇H₁₈BrFO₂
MW: 353.2260 g/mol
Storage Temperature: Ambient
CAS Number: 3002427-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR