1-(6-Bromo-2-fluoro-3-propoxyphenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

314264-1G 314264-500MG 314264-5G
77468-928EA 415.09 USD
77468-928 77481-610 77491-996
1-(6-Bromo-2-fluoro-3-propoxyphenyl)prop-2-en-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-propoxyphenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₁₂H₁₄BrFO₂
MW: 289.1407 g/mol
Storage Temperature: Ambient
CAS Number: 3002529-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR