(6-Bromo-2-fluoro-3-propoxyphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

314262-1G 314262-500MG 314262-5G
77468-924EA 415.09 USD
77468-924 77481-606 77491-992
(6-Bromo-2-fluoro-3-propoxyphenyl)triethylsilane ≥95%
(6-Bromo-2-fluoro-3-propoxyphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₅H₂₄BrFOSi
MW: 347.3383 g/mol
Storage Temperature: Ambient
CAS Number: 3002541-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR