(6-Bromo-2-fluoro-3-propoxyphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

314260-1G 314260-500MG 314260-5G
77468-920EA 415.09 USD
77468-920 77481-602 77491-988
(6-Bromo-2-fluoro-3-propoxyphenyl)(cyclopropyl)methanol ≥95%
(6-Bromo-2-fluoro-3-propoxyphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₆BrFO₂
MW: 303.1673 g/mol
Storage Temperature: Ambient
CAS Number: 3002434-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR