(6-Bromo-2-fluoro-3-propoxyphenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

314256-1G 314256-500MG 314256-5G
77468-916EA 415.09 USD
77468-916 77481-598 77491-984
(6-Bromo-2-fluoro-3-propoxyphenyl)(4-ethylphenyl)methanol ≥95%
(6-Bromo-2-fluoro-3-propoxyphenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₈H₂₀BrFO₂
MW: 367.2526 g/mol
Storage Temperature: Ambient
CAS Number: 3002429-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR