(6-Bromo-2-fluoro-3-methoxyphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

314234-1G 314234-500MG 314234-5G
77468-912EA 415.09 USD
77468-912 77481-594 77491-980
(6-Bromo-2-fluoro-3-methoxyphenyl)(cyclohexyl)methanol ≥95%
(6-Bromo-2-fluoro-3-methoxyphenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₈BrFO₂
MW: 317.1939 g/mol
Storage Temperature: Ambient
CAS Number: 3002529-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR