(6-Bromo-2-fluoro-3-methoxyphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

314232-1G 314232-500MG 314232-5G
77468-908EA 415.09 USD
77468-908 77481-590 77491-976
(6-Bromo-2-fluoro-3-methoxyphenyl)triethylsilane ≥95%
(6-Bromo-2-fluoro-3-methoxyphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₂₀BrFOSi
MW: 319.2852 g/mol
Storage Temperature: Ambient
CAS Number: 3002467-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR