(6-Bromo-2-fluoro-3-methoxyphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

314227-1G 314227-500MG 314227-5G
77468-900EA 415.09 USD
77468-900 77481-582 77491-968
(6-Bromo-2-fluoro-3-methoxyphenyl)(cyclopropyl)methanol ≥95%
(6-Bromo-2-fluoro-3-methoxyphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₂BrFO₂
MW: 275.1141 g/mol
Storage Temperature: Ambient
CAS Number: 3002473-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR