(6-Bromo-2-fluoro-3-methoxyphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

314225-1G 314225-500MG 314225-5G
77468-896EA 415.09 USD
77468-896 77481-578 77491-964
(6-Bromo-2-fluoro-3-methoxyphenyl)(ethyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-methoxyphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀BrFOS
Storage Temperature: Ambient
CAS Number: 3002427-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR