(6-Bromo-2-fluoro-3-methoxyphenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

314221-1G 314221-500MG 314221-5G
77468-892EA 415.09 USD
77468-892 77481-574 77491-960
(6-Bromo-2-fluoro-3-methoxyphenyl)(4-ethylphenyl)methanol ≥95%
(6-Bromo-2-fluoro-3-methoxyphenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₆BrFO₂
MW: 339.1994 g/mol
Storage Temperature: Ambient
CAS Number: 3002527-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR