3-(Dimethylamino)-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31406-1G 31406-500MG 31406-5G
77468-888EA 195.7 USD
77468-888 77481-570 77491-956
3-(Dimethylamino)-5-fluorobenzaldehyde ≥97%
3-(Dimethylamino)-5-fluorobenzaldehyde

Precaución: For research use only

Formula: C₉H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD29121746
CAS Number: 1289041-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR