1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

313170-1G 313170-500MG 313170-5G
77468-884EA 415.09 USD
77468-884 77481-566 77491-952
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2-methylpropan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₁BrF₄O
MW: 315.1020 g/mol
Storage Temperature: Ambient
CAS Number: 3002503-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR