1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥97%

Supplier: AOBChem USA

Warning

313165-1G 313165-500MG 313165-5G
77468-876EA 370.8 USD
77468-876 77481-558 77491-944
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥97%
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₀H₂BrF₉O
Storage Temperature: Ambient
CAS Number: 3002497-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR