1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

313162-1G 313162-500MG 313162-5G
77468-872EA 415.09 USD
77468-872 77481-554 77491-940
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₃BrF₄O
MW: 329.1286 g/mol
Storage Temperature: Ambient
CAS Number: 3002435-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR