2-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

313158-1G 313158-500MG 313158-5G
77468-864EA 415.09 USD
77468-864 77481-546 77491-932
2-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₉BrF₄O
MW: 301.0754 g/mol
Storage Temperature: Ambient
CAS Number: 3002503-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR