Ethyl(4-methylnaphthalen-1-yl)sulfane ≥95%

Supplier: AOBChem USA

Warning

313129-1G 313129-500MG 313129-5G
77468-860EA 415.09 USD
77468-860 77481-542 77491-928
Ethyl(4-methylnaphthalen-1-yl)sulfane ≥95%
Ethyl(4-methylnaphthalen-1-yl)sulfane

Precaución: For research use only

Formula: C₁₃H₁₄S
Storage Temperature: Ambient
CAS Number: 3002503-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR