1-(4-Methylnaphthyl) trifluoromethyl ketone ≥96%

Supplier: AOBChem USA

Warning

313126-1G 313126-500MG 313126-5G
77468-856EA 266.77 USD
77468-856 77481-538 77491-924
1-(4-Methylnaphthyl) trifluoromethyl ketone ≥96%
1-(4-Methylnaphthyl) trifluoromethyl ketone

Precaución: For research use only

Formula: C₁₃H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD01320010
CAS Number: 886370-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR