1-([1,1'-biphenyl]-2-yl)-2-chloro-2,2-difluoroethanone 95

Supplier: AOBChem USA

Warning

313119-1G 313119-500MG 313119-5G
77468-852EA 284.28 USD
77468-852 77481-534 77491-920
1-([1,1'-biphenyl]-2-yl)-2-chloro-2,2-difluoroethanone 95
1-([1,1'-biphenyl]-2-yl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₄H₉ClF₂O
MW: 266.6704 g/mol
Storage Temperature: Ambient
CAS Number: 1989793-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR