4-Chloro-3-fluoro-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

31250-1G 31250-500MG 31250-5G
77468-844EA 346.08 USD
77468-844 77481-526 77491-912
4-Chloro-3-fluoro-2-methoxyphenylboronic acid ≥97%
4-Chloro-3-fluoro-2-methoxyphenylboronic acid

Precaución: For research use only

Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414971
CAS Number: 2121515-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR