5-Chloro-2-fluoro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

31193-1G 31193-500MG 31193-5G
77468-840EA 192.61 USD
77468-840 77481-522 77491-906
5-Chloro-2-fluoro-4-methoxybenzoic acid ≥97%
5-Chloro-2-fluoro-4-methoxybenzoic acid

Precaución: For research use only

Formula: C₈H₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD18441714
CAS Number: 211172-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR