5-Chloro-2-fluoro-4-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

31190-1G 31190-500MG 31190-5G
77468-836EA 457.32 USD
77468-836 77481-518 77491-902
5-Chloro-2-fluoro-4-methoxyphenol ≥97%
5-Chloro-2-fluoro-4-methoxyphenol

Precaución: For research use only

Formula: C₇H₆ClFO₂
MW: 176.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414652
CAS Number: 1394953-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR