3-[(3-Methylbenzyl)oxy]benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-[(3-Methylphenyl)methoxy]benzaldehyde

311635-1G 311635-500MG 311635-5G
77468-832EA 85.49 USD
77468-832 77481-514 77491-896
3-[(3-Methylbenzyl)oxy]benzaldehyde ≥95%
3-[(3-Methylbenzyl)oxy]benzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03422455
CAS Number: 590354-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR