B-[4-(Cyclopropylmethoxy)-2-methylphenyl]boronic acid ≥95%

Supplier: AOBChem USA

Warning

310777-1G 310777-500MG 310777-5G
77468-828EA 785.89 USD
77468-828 77481-510 77491-894
B-[4-(Cyclopropylmethoxy)-2-methylphenyl]boronic acid ≥95%
B-[4-(Cyclopropylmethoxy)-2-methylphenyl]boronic acid

Precaución: For research use only

Formula: C₁₁H₁₅BO₃
Storage Temperature: Ambient
CAS Number: 2246652-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR